Order Entry
Slovakia
ContactUsLinkComponent
SEED TESTING PAPER SHEETS 200 X 350 MM, 430 G/M, DARK BLUE 1 * 100 items
Catalog # BINZ2.383.200350
Supplier:  Ahlstrom-Munksjö
SEED TESTING PAPER SHEETS 200 X 350 MM, 430 G/M, DARK BLUE 1 * 100 items
Catalog # BINZ2.383.200350
Supplier:  Ahlstrom-Munksjö
Frequently Bought Together