Order Entry
Slovakia
ContactUsLinkComponent
ROCK1(THR455+SER456) POLYCLONAL ANTIBODY, ALEXA FLUOR® 750 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPL ICATION: IF(IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-10239R-A750
Supplier:  BIOSS INC
ROCK1(THR455+SER456) POLYCLONAL ANTIBODY, ALEXA FLUOR® 750 CONJUGATED, HOST: RABBIT, CROSS REACTIVE SPECIES: HUMAN, MOUSE, RAT, APPL ICATION: IF(IHC-P), CONCENTRATION: 1UG/UL, VOLUME: 100UL 1 * 100 µl
Catalog # BOSSBS-10239R-A750
Supplier:  BIOSS INC