Order Entry
Slovakia
ContactUsLinkComponent
Mouse BTLA, His Tag (BTA-M52E2) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession Q32MV9). 1 * 1 mg
Catalog # ACRBBTA-M52E2-1MG
Supplier:  ACRO Biosystems
Mouse BTLA, His Tag (BTA-M52E2) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession Q32MV9). 1 * 1 mg
Catalog # ACRBBTA-M52E2-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together