Order Entry
Slovakia
ContactUsLinkComponent
g-trap protein g are prepacked ready to use columns for affinity chromatography. the column is packed with resin, which is covalent ly coupled to recombinant protein g. the g-trap columns are made of biocompatible polypropylene, which does not int 1 * 5 mL
Catalog # GENO786-1035
Supplier:  G BIOSCIENCES
g-trap protein g are prepacked ready to use columns for affinity chromatography. the column is packed with resin, which is covalent ly coupled to recombinant protein g. the g-trap columns are made of biocompatible polypropylene, which does not int 1 * 5 mL
Catalog # GENO786-1035
Supplier:  G BIOSCIENCES