Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Knitted caps, B013

Knitted caps, B013

Knitted caps, Yellow
Catalog # PRTWB013YER
Supplier:   Portwest
Knitted caps, B013
Catalog # PRTWB013YER
Supplier:  Portwest
Knitted caps, B013
Knitted caps, B013

Knitted caps, Yellow

Price
7,40 €
Log in for availability
1 * 1 items
Product Details & Documents

These caps are made from 100% acrylic knit with a 40 g Insulatex™ lining. Ideal for really cold days.

  • Insulatex™ lining ensures that the wearer remains warm
  • Available in a choice of colours
Specifications
  • Type:
    Knitted caps
  • Colour:
    Yellow
  • Seq:
    05
  • Cat. No.:
    PRTWB013YER
  • Risk area:
    Cold
  • Model:
    B013
  • Garment application:
    Outdoor
  • Gender:
    Unisex
  • Fabric:
    100% acrylic knit with Insulatex™ lining