0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Slovakia
ContactUsLinkComponent
Mouse Anti-Melanoma, gp100 (pMel17, Melanocyte Protein PMEL, Melanocytes Lineage-specific Antigen GP100) (APC) 1 * 100 µl
USBI138569-AF-APC
:
UNITED BIOCHEMICALS
Mouse Anti-Melanoma, gp100 (pMel17, Melanocyte Protein PMEL, Melanocytes Lineage-specific Antigen GP100) (APC) 1 * 100 µl
USBI138569-AF-APC
:
UNITED BIOCHEMICALS
About USBiological
Frequently Bought Together
Sealing film for manual application, Parafilm® M
: HEATHROW
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
VWR® NITRILE LIGHT, Nitrile Gloves
: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
: VWR Collection
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Nitrile gloves, Kimtech™ Purple Nitrile™
: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR® Facial Tissues
: VWR Collection
VWR®, Nitrile Gloves
: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals