Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Ballotini solid soda glassballs 0.0-0.02mm dia Grd 5209 1 * 1 kg
Catalog # LINR5209
Supplier:
SIGMUND LINDNER
Expand image
Ballotini solid soda glassballs 0.0-0.02mm dia Grd 5209 1 * 1 kg
Catalog # LINR5209
Supplier:
SIGMUND LINDNER
Frequently Bought Together
VWR®, Glass Stoppers for Standard Ground Joints
Supplier: VWR Collection
Laboratory bottles, narrow neck, with screw cap, DURAN®
Supplier: DWK LIFE SCIENCES
VWR®, Beakers, Low Form, Heavy-Duty
Supplier: VWR Collection
VWR® Flasks, Round Bottom, with Standard Ground joint
Supplier: VWR Collection
Toluene ≥99.90%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
VWR®, Beakers, Tall Form
Supplier: VWR Collection
VWR®, Beakers, Low Form
Supplier: VWR Collection
VWR®, Pasteur Pipettes
Supplier: VWR Collection
VWR®, Laboratory Bottles, Narrow Neck, with Screw Cap
Supplier: VWR Collection
Screw caps for laboratory bottles, DURAN®
Supplier: DWK LIFE SCIENCES