Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Accessories for Falcon® pipette controller

Accessories for Falcon® pipette controller

Universal power supply (Input: 100 - 250 V, Output: 9 V)
Catalog # BDAA357489
   Low Manufacturing Impact
Supplier:   Corning
Accessories for Falcon® pipette controller
Catalog # BDAA357489
Supplier:  Corning
   Low Manufacturing Impact
Accessories for Falcon® pipette controller
Accessories for Falcon® pipette controller

Universal power supply (Input: 100 - 250 V, Output: 9 V)

Price
43,16 €
Log in for availability
1 * 1 items
Product Details & Documents
Specifications
  • Description:
    Universal power supply (Input: 100 - 250 V, Output: 9 V)
  • Seq:
    17
  • Cat. No.:
    BDAA357489
  • Environmental certification:
    I-RECs, ISO 14001 - Warsaw
  • Environmentally Preferable:
    Low Manufacturing Impact