Order Entry
Slovakia
ContactUsLinkComponent
This product is prepared by labeling high quality goat anti-rabbit IgG (H+L) with alkaline phosphatase (AP). AP conjugates could be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications. 1 * 100 µl
Catalog # 20465-100UL
Supplier:  BIOTIUM
This product is prepared by labeling high quality goat anti-rabbit IgG (H+L) with alkaline phosphatase (AP). AP conjugates could be used for Western Blot, ELISA, immunohistochemistry and other standard immunoassay applications. 1 * 100 µl
Catalog # 20465-100UL
Supplier:  BIOTIUM
Frequently Bought Together