An activator of Ca2+-sensitive K+ channels (EC50 = 490 µM in T84 human carcinoma cells) 1 * 5 mg
Catalog # CAYM14575-5
Supplier: CAYMAN
An activator of Ca2+-sensitive K+ channels (EC50 = 490 µM in T84 human carcinoma cells) 1 * 5 mg
Catalog # CAYM14575-5
Supplier: CAYMAN
Frequently Bought Together
96-Well clear polystyrene microplates, Corning®
Supplier: Corning
VWR®, Cryoboxes, Half Size, Cardboard
Supplier: VWR Collection
VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
VWR®, Square Bottles, PET
Supplier: LAB3 OWN PRODUCTION
VWR®, Cell Strainers
Supplier: VWR Collection
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Supplier: EPPENDORF
Multiwell cell culture plates, Falcon®
Supplier: Corning
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Supplier: EPPENDORF
Serological pipettes, Stripette™
Supplier: Corning
VWR®, Cryoboxes, Cardboard, 133×133 mm
Supplier: VWR Collection
Cryoboxes, B69/B50/B80/B99-Series
Supplier: GLW STORING SYSTEMS
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, PCR/qPCR Plates, 96-Wells, Plastic
Supplier: VWR Collection
Serological pipettes
Supplier: Greiner Bio-One
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
VWR®, Spreaders, L-Shaped, Standard Line
Supplier: VWR Collection
Cell culture storage bottles, polystyrene, Corning®
Supplier: Corning