Order Entry
Slovakia
ContactUsLinkComponent
An inhibitor of the DNA methyltransferases DNMT1, DNMT3A, and DNMT3B (IC50s = 12.5, 8, and 7.5 M, respectively, with Poly(dI-dC) as the substrate) 1 * 5 mg
Catalog # CAYM11165-5
Supplier:  CAYMAN
An inhibitor of the DNA methyltransferases DNMT1, DNMT3A, and DNMT3B (IC50s = 12.5, 8, and 7.5 M, respectively, with Poly(dI-dC) as the substrate) 1 * 5 mg
Catalog # CAYM11165-5
Supplier:  CAYMAN
Frequently Bought Together