Order Entry
Slovakia
ContactUsLinkComponent
A selective autotaxin inhibitor (IC50 = 5.7 nM) that has been shown to dose-dependently block thrombin-induced LPA secretion in plat elets 1 * 10 mg
Catalog # CAYM11034-10
Supplier:  CAYMAN
A selective autotaxin inhibitor (IC50 = 5.7 nM) that has been shown to dose-dependently block thrombin-induced LPA secretion in plat elets 1 * 10 mg
Catalog # CAYM11034-10
Supplier:  CAYMAN
Frequently Bought Together