Order Entry
Slovakia
ContactUsLinkComponent
30µG/G AG, AL, B, BA, CA, CD, CR, CU, FE, K, MG, MN, MO, NA, NI, P, PB, SI, SN, TI, V, ZN IN HYDROCARBON OIL 1 * 1 items
Catalog # PROOGV21-K-30-200G
Supplier:  PROMOCHEM
30µG/G AG, AL, B, BA, CA, CD, CR, CU, FE, K, MG, MN, MO, NA, NI, P, PB, SI, SN, TI, V, ZN IN HYDROCARBON OIL 1 * 1 items
Catalog # PROOGV21-K-30-200G
Supplier:  PROMOCHEM
Frequently Bought Together