Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Prihlásiť sa
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
1-[4-(1,1-DIMETHYLETHYL)PHENYL]-4-(4-HYDROXYPIPERIDIN-1-YL)BUTAN-1-ONE 1 * 100 mg
Catalog # PROOMM0468.05
Dodávateľ:
PROMOCHEM
Expand image
1-[4-(1,1-DIMETHYLETHYL)PHENYL]-4-(4-HYDROXYPIPERIDIN-1-YL)BUTAN-1-ONE 1 * 100 mg
Catalog # PROOMM0468.05
Dodávateľ:
PROMOCHEM
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Dodávateľ: Merck
Potassium hydrogen phthalate, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
Dodávateľ: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Dodávateľ: VWR Chemicals
VWR®, Conductivity standard solutions
Dodávateľ: VWR Chemicals
pH buffers, DuraCal
Dodávateľ: HAMILTON BONADUZ
Buffer solutions in sachets, Certipur®, Supelco®
Dodávateľ: Merck
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Dodávateľ: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Dodávateľ: Merck
Pure water density standard, Supelco®
Dodávateľ: Merck