Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

Potassium iodide 99+% for analysis

Glass bottle, 2,5 kg
Catalog # ACRO373650025
Health
Potassium iodide 99+% for analysis
Catalog # ACRO373650025
CAS Number:  7681-11-0
Potassium iodide 99+% for analysis
Potassium iodide 99+% for analysis

Glass bottle, 2,5 kg

CAS Number:  7681-11-0
Price
896,42 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
Synonyms:  Dipotassium diiodide, Kali iodide
  • CAS Number:7681-11-0
  • Formula:Kl
  • MDL Number:MFCD00011405
  • Molecular Weight:166 g/mol
  • Merck Index:13,07727
  • Boiling Pt:1330 °C (760 mmHg)
  • Melting Pt:680 °C
  • Density:3,13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-659-4
Specifications

Specification Test Results

  • Appearance:
    Colorless to white Crystalline powder or crystals
  • Titration Argentometric:
    ≥99.0 % (on dry substance)
  • Loss on drying:
    ≤0.2 % (150°C, 6 hrs)
  • pH:
    6.0 to 9.2 (5 % at 25°C)
  • Alkalinity:
    ≤0.022 % (as KOH)
  • Iodate (IO3):
    ≤2 ppm
  • Sulfate (SO4):
    ≤50 ppm
  • Iron (Fe):
    ≤5 ppm
  • Heavy metals (as Pb):
    ≤10 ppm
  • Nitrogen compounds (as N):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.