Order Entry
Slovakia
ContactUsLinkComponent
PEQSTAR 96X UNIVERSAL GRADIENTCycler with USB & network capabilityUniversal block for 96 x 0.2 ml tubes,96 well PCR plates or 48 x 0.5 ml tubeswith flat lid. 1 * 1 item
Catalog # 732-2887
Supplier:  VWR Peqlab
undefined
PEQSTAR 96X UNIVERSAL GRADIENTCycler with USB & network capabilityUniversal block for 96 x 0.2 ml tubes,96 well PCR plates or 48 x 0.5 ml tubeswith flat lid. 1 * 1 item
Catalog # 732-2887
Supplier:  VWR Peqlab
Frequently Bought Together