M-GREEN AGAR 1 * 500 g
SCAR01-633-500
: SCHARLAB
Frequently Bought Together
Isobutanol 99+% for HPLC
: Thermo Fisher Scientific
Ethyl Acetate (BP, Ph. Eur.) pharma grade 1 * 200 L
: PanReac AppliChem
Isoamyl alcohol 98%, pure
: Thermo Fisher Scientific
1-Propanol ≥99%, GPR RECTAPUR®
: VWR Chemicals
3-Nitrobenzyl alcohol, Supelco®
: Merck
1-Pentanol ≥98%
: Thermo Fisher Scientific
Xylene (mixture of isomers)
: LACH NER
Water ACS
: Thermo Fisher Scientific
Dimethyl sulphoxide 99.8+% for HPLC
: Thermo Fisher Scientific
Liquid for manometer with sloping tube
: VWR Chemicals
n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals