Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

Imipenem monohydrate

100 mg
Katalógové číslo CAYM16039-100
Dodávateľ:   CAYMAN
Imipenem monohydrate
Katalógové číslo CAYM16039-100
Dodávateľ:  CAYMAN
Číslo CAS:  74431-23-5
Imipenem monohydrate

100 mg

Prejsť na:
Číslo CAS:  74431-23-5
Price
567,61 €
Prihlásiť sa pre dostupnosť
1 * 100 mg
Product Details & Documents

Imipenem is an intravenous β-lactam antibiotic effective against a wide range of Gram-positive and Gram-negative bacteria (MICs range from 0,5 - 2 µg/ml for E. cloacae and P. aeruginosa), including several multi-drug resistant bacterial species. Imipenem acts by interfering with bacterial cell wall synthesis.

  • A crystalline solid
Synonymá:  (5R,6S)-3-((2-Formimidamidoethyl)thio)-6-((R)-1-hydroxyethyl)-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylic acid hydrate
  • Číslo CAS:74431-23-5
  • Vzorec:C₁₂H₁₇N₃O₄S
  • MDL číslo:MFCD09753321
  • Molecular Weight:317.36 g/mol
  • Teplota pre skladovanie:Freezer
Špecifikácie

Špecifikácie

  • Pk:
    100 mg

Špecifikácie

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.