Temperature indicating kit
Thermax chill checker, type 8, +20 °C
Catalog # KLEI1216168
Supplier: KLEINFELD
Jump to:
Product Details & Documents
These Thermax chill checker include eight different measurement points over a temperature range from −17 to +20 °C. Activation takes place after cooling by pressing in the center. When the temperature is exceeded, it changes color purple or blue.
Specifications
- Max. temp.:+20 °C
- Description:Thermax chill checker, type 8
- Seq:03