Order Entry
Slovakia
ContactUsLinkComponent
The Liberty Blue System is an automated microwave peptide synthesizer designed for Fmoc solid phase peptide synthesis from 0.025 - 1mmol scale using reaction vessels ranging from 10-125mL volume. The Liberty Blue uses a Discover single mode microwa 1 * 1 items
Catalog # CEMA925602
Supplier:  CEM
The Liberty Blue System is an automated microwave peptide synthesizer designed for Fmoc solid phase peptide synthesis from 0.025 - 1mmol scale using reaction vessels ranging from 10-125mL volume. The Liberty Blue uses a Discover single mode microwa 1 * 1 items
Catalog # CEMA925602
Supplier:  CEM
Frequently Bought Together