Order Entry
Slovakia
ContactUsLinkComponent
qPCR detection of Western equine encephalomyelitis virus (WEEV). Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-WEEV
Supplier:  PRIMERDESIGN
qPCR detection of Western equine encephalomyelitis virus (WEEV). Contains: Primer and probe, RT target specific primers (RNA viruses only), Standard curve, Endogenous control, extraction control 1 * 150 Tests
Catalog # PATH-WEEV
Supplier:  PRIMERDESIGN
Frequently Bought Together