Order Entry
Slovakia
ContactUsLinkComponent
qPCR detection of Hand, foot and mouth disease. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA g enome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HFMD-
Supplier:  PRIMERDESIGN
qPCR detection of Hand, foot and mouth disease. Contains: Primer and probe mix, Reverse Transcription target specific primers (RNA g enome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-HFMD-
Supplier:  PRIMERDESIGN
Frequently Bought Together