Order Entry
Slovakia
ContactUsLinkComponent
qPCR detection of Infectious Hematopoietic Necrosis Virus. Contains: Primer and probe mix, Reverse Transcription target specific pri mers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-IHNV-
Supplier:  PRIMERDESIGN
qPCR detection of Infectious Hematopoietic Necrosis Virus. Contains: Primer and probe mix, Reverse Transcription target specific pri mers (RNA genome viruses only), Copy number standard curve 1 * 150 Tests
Catalog # PATH-STD-IHNV-
Supplier:  PRIMERDESIGN
Frequently Bought Together