Order Entry
Slovakia
ContactUsLinkComponent
GRO-Beta Viral Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 70 amino acids and having a molecular mass of 7.9 kDa. The CXCL2 is purified by proprietary chromatographic techniques. 1 * 50 µG
Catalog # BIRBORB80134-50
Dodávateľ:  BIORBYT
GRO-Beta Viral Recombinant produced in E.coli is a single,non-glycosylated, polypeptide chain containing 70 amino acids and having a molecular mass of 7.9 kDa. The CXCL2 is purified by proprietary chromatographic techniques. 1 * 50 µG
Catalog # BIRBORB80134-50
Dodávateľ:  BIORBYT
Súvisiace informácie
Frequently Bought Together