Order Entry
Slovakia
ContactUsLinkComponent
Polyvalent dengue antigen contains dengue antigen subtypes. The immunoreactive peptide span from the viral envelope to the non-funct ional region. The polyvalent dengue antigen consists of 20% recombinant antigen from each of the dengue subt 1 * 500 µG
Catalog # BIRBORB81813-500
Supplier:  BIORBYT
Polyvalent dengue antigen contains dengue antigen subtypes. The immunoreactive peptide span from the viral envelope to the non-funct ional region. The polyvalent dengue antigen consists of 20% recombinant antigen from each of the dengue subt 1 * 500 µG
Catalog # BIRBORB81813-500
Supplier:  BIORBYT