Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Laboratory coats, PP
Laboratory coats, PP
White, 3XL
Catalog # 113-8132
Laboratory coats, PP
Catalog # 113-8132
Laboratory coats, PP
Laboratory coats, PP
White, 3XL
Price
97,82 €
Log in for availability
1 * 50 items
Product Details & Documents
These disposable laboratory coats made from PP are ideal for non critical applications.
  • Elasticated wrist
  • Press stud fastening (four snaps)
  • Two pockets below the waist
Weight: 30 g/m²
Colour: White
Documents
Specifications
  • Clothing Size:
    3XL
  • Colour:
    White
  • Seq:
    05
  • Cat. No.:
    113-8132
  • Garment application:
    Laboratory
  • Gender:
    Unisex
  • Pockets:
    With pockets
  • Closure type:
    Press studs / snaps
  • Weight:
    30 g/m²