Order Entry
Slovakia
ContactUsLinkComponent
HayeSep A, 80/100, preconditioned, 41.28 mm coil, 5.58 feet (1.7 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Katalógové číslo HEWLG3591-81210
undefined
HayeSep A, 80/100, preconditioned, 41.28 mm coil, 5.58 feet (1.7 m) x 1/8 inch, 2.1 mm, UltiMetal, packed GC column, for Large Valve Oven 1 * 1 items
Katalógové číslo HEWLG3591-81210
Frequently Bought Together