Order Entry
Slovakia
ContactUsLinkComponent
260 x USP System Suitability Set consisting of 1 x 40ml vial of Reagent Water (RTOCW), Standard Solution (RTOCRs) and Suitability So lution (RTOCRss) Delivered at 2 weekly intervals 1 * 40 mL
Catalog # REGERTOCUSP260
Supplier:  REAGECON
260 x USP System Suitability Set consisting of 1 x 40ml vial of Reagent Water (RTOCW), Standard Solution (RTOCRs) and Suitability So lution (RTOCRss) Delivered at 2 weekly intervals 1 * 40 mL
Catalog # REGERTOCUSP260
Supplier:  REAGECON
Frequently Bought Together