Order Entry
Slovakia
ContactUsLinkComponent
Gas25h2s,100co,2.2%ch4,18%o2,n2 34l 4ga18x22w25h100-34 1 * 1 items
Catalog # BWTE4GA18X22W25H34
Supplier:  BW TECHNOLOGIES
Gas25h2s,100co,2.2%ch4,18%o2,n2 34l 4ga18x22w25h100-34 1 * 1 items
Catalog # BWTE4GA18X22W25H34
Supplier:  BW TECHNOLOGIES
Frequently Bought Together