Dropping bottles, glass
20 ml, DIN 18
Catalog # HECH41314020
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Made of amber glass.
- Good chemical and physical properties
- For use in applications that do not involve high thermal stresses
- Amber coloured to protect light sensitive substances
Dropping bottles without cap, with thread DIN 18.
Informace pro objednávání: Pipette kit and screw cap must be ordered seperately.
Specifications
- Thread:DIN 18
- Capacity:20 ml
- Seq:02
Accessories
Product Information
- Catalog #AvailabilityPrice
- Pipette kit, red, for 5 ml dropping bottlesPipette kit, red, for 5 ml dropping bottlesHECH41315006Log in for availability1 * 100 items - 68,64 €
Product Information
- Catalog #AvailabilityPrice
- Pipette kit, blue, for 20 ml dropping bottlesPipette kit, blue, for 20 ml dropping bottlesHECH41315020Log in for availability1 * 100 items - 66,58 €
Product Information
- Catalog #AvailabilityPrice
- Pipette kit, blue, for 30 ml dropping bottlesPipette kit, blue, for 30 ml dropping bottlesHECH41315030Log in for availability1 * 100 items - 68,64 €
Product Information
- Catalog #AvailabilityPrice
- Pipette kit, red, for 30 ml dropping bottlesPipette kit, red, for 30 ml dropping bottlesHECH41315031Log in for availability1 * 100 items - 68,64 €
Product Information
- Catalog #AvailabilityPrice
- Pipette kit, red, for 100 ml dropping bottlesPipette kit, red, for 100 ml dropping bottlesHECH41315061Log in for availability1 * 100 items - 69,87 €
Product Information
- Catalog #AvailabilityPrice
- Pipette kit, red, for 20 ml dropping bottlesPipette kit, red, for 20 ml dropping bottlesHECH41315021Log in for availability1 * 100 items - 66,58 €
Product Information
- Catalog #AvailabilityPrice
- Pipette kit, red, for 10 ml dropping bottlesPipette kit, red, for 10 ml dropping bottlesHECH41315011Log in for availability1 * 100 items - 66,58 €
Product Information
- Catalog #AvailabilityPrice
- Pipette kit, red, for 50 ml dropping bottlesPipette kit, red, for 50 ml dropping bottlesHECH41315051Log in for availability1 * 100 items - 67,82 €
Product Information
- Catalog #AvailabilityPrice
- Pipette kit, blue, for 50 ml dropping bottlesPipette kit, blue, for 50 ml dropping bottlesHECH41315650Log in for availability1 * 100 items - 48,09 €