Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Volumetric flasks, class A

Volumetric flasks, class A

25 ml, 10/19, 0,040 ± ml
Catalog # GLAS129.234.03
Supplier:   GLASSCO
Volumetric flasks, class A
Catalog # GLAS129.234.03
Supplier:  GLASSCO
Volumetric flasks, class A
Volumetric flasks, class A

25 ml, 10/19, 0,040 ± ml

Price
7,60 €
Log in for availability
1 * 1 items
Product Details & Documents

Borosilicate glass 3.3, clear, with hollow glass stopper.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Autoclavable at 121 °C

Certifications: According to DIN 12664 and ISO 1042.

Specifications
  • NS:
    10/19
  • Capacity:
    25 ml
  • Seq:
    04
  • Tolerance:
    0,040 ± ml