Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Volumetric flasks, class A

Volumetric flasks, class A

10 ml, 10/19, 0,025 ± ml
Catalog # GLAS129.234.02
Supplier:   GLASSCO
Volumetric flasks, class A
Catalog # GLAS129.234.02
Supplier:  GLASSCO
Volumetric flasks, class A
Volumetric flasks, class A

10 ml, 10/19, 0,025 ± ml

Price
6,82 €
Log in for availability
1 * 1 items
Product Details & Documents

Borosilicate glass 3.3, clear, with hollow glass stopper.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • Autoclavable at 121 °C

Certifications: According to DIN 12664 and ISO 1042.

Specifications
  • NS:
    10/19
  • Capacity:
    10 ml
  • Seq:
    02
  • Tolerance:
    0,025 ± ml