Potassium permangamate N/10 1 * 1 ST
Catalog # LACH61022050P0000
Supplier: LACH NER
Frequently Bought Together
PETRI DISH 60X14MM 1 * 960 ST
Supplier: GAMA GROUP
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Hypodermic needles, HENKE-JECT® for single use
Supplier: HENKE SASS WOLF
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
pH electrodes, SenTix® PLUS
Supplier: WTW
Sodium hypochlorite 6…14% (active chlorine) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Sodium chloride 99.5-100.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals