Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Acetone

Acetone

1 L
Catalog # LACH20001CT0M1000
Flammable
Irritant
   Safer Human and Environmental Health
Supplier:   LACH NER
Acetone
Catalog # LACH20001CT0M1000
Supplier:  LACH NER
   Safer Human and Environmental Health
CAS Number:  67-64-1
Acetone
CAS Number:  67-64-1
Price
9,74 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specifications

  • Pk:
    1 L
  • Cat. No.:
    LACH20001CT0M1000
  • Environmental certification:
    Chem21 Solvent Selection Guide recommended after discussion
  • Environmentally Preferable:
    Safer Human and Environmental Health

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.