Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Acetone

Acetone

Glass bottle, 7.900 g
Catalog # LACH20001AT0G7900
Flammable
Irritant
Supplier:   LACH NER
Acetone
Catalog # LACH20001AT0G7900
Supplier:  LACH NER
CAS Number:  67-64-1
Acetone
Acetone

Glass bottle, 7.900 g

CAS Number:  67-64-1
Price
62,06 €
Log in for availability
1 * 7.900 g
Product Details & Documents
Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:13,00067
  • Boiling Pt:56.2 °C (1013 hPa)
  • Melting Pt:–95.4 °C
  • Density:0.792 g/cm³ (20 °C)
  • Flash Pt:<–20 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    7.900 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.