[CS]PUFR AMONIAKÃLNÃ PH 10 1 * 1 L
Catalog # LACH64001000M10001
Supplier: LACH NER
Frequently Bought Together
SOYASAPOGENOL B 1 * 5 mg
Supplier: Merck
Automatic burettes, Pellet pattern, lateral stopcock
Supplier: HIRSCHMANN
Tantalum standard solution, for ICP, in 2% HF, CRM, VHG
Supplier: PROMOCHEM
Iohexol, Reference Standards
Supplier: USP
2,3-Dimercapto-1-propanol 97.0-100.5%
Supplier: Thermo Fisher Scientific
Raspberry Alcohol, Reference Standard
Supplier: USP
Analytical Volumetric Solution Zinc Sulphate 0.1N 0.1M 1 * 1 L
Supplier: REAGECON
Supelco® PESTANAL® Organic Reference Standard, Hexazinone
Supplier: Merck
OXALATE IC-STANDARD 1 * 100 mL
Supplier: REAGECON
Iridium standard solution, 1000 mg/L Ir in 20% hydrochloric acid (from IrCl₃), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Methyl enantate
Supplier: EHRENSTORFER
TRIGLYCINE 1 * 100 mg
Supplier: Merck
Ibuprofen, Supelco®
Supplier: Merck