Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Blower foot switch (foot pedal and power cord), provides a hands-free means to temporarily shut off power to the blower when obtaining sensitive balance readings or manipulating fine powders
Catalog # LBCP5441300
Supplier:   LABCONCO
Accessories for glove boxes, Protector®
Catalog # LBCP5441300
Supplier:  LABCONCO
Accessories for glove boxes, Protector®
Accessories for glove boxes, Protector®
Blower foot switch (foot pedal and power cord), provides a hands-free means to temporarily shut off power to the blower when obtaining sensitive balance readings or manipulating fine powders
Price
759,15 €
Log in for availability
1 * 1 items
Product Details & Documents
Documents
Specifications
  • Description:
    Blower foot switch (foot pedal and power cord), provides a hands-free means to temporarily shut off power to the blower when obtaining sensitive balance readings or manipulating fine powders
  • Seq:
    14
Odporúčania
Odporúčania budú personalizované na základe vašich nákupných preferencií len v prípade, že ste udelili súhlas aktiváciou možnosti "Vylepšiť môj nákupný zážitok"na stránke "Osobné údaje – Nastavenia ochrany osobných údajov“
V opačnom prípade dostanete všeobecné odporúčania.