Laboratory chair neon, upholstery made of pu-foam, blue, impervious to mild acids and alkalines, seat inclination adjustment, seat h eight adjustment with a gasspring from 450-620 mm, 5-star base made of aluminium, with glides, with foot ring 1 * 1 items
Catalog # BIMO57120013279525
Supplier: BIMOS SITZTECHNIK
Frequently Bought Together