Cyclohexane
Glass bottle, 2,5 L
Catalog # SCARCI00352500
Supplier: SCHARLAB
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Boiling Pt:81 °C (1013 hPa)
- Melting Pt:7 °C
- Density:0.7785 g/cm³ (20 °C)
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle
- Pk:2,5 L



