Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

(±)-7-HDoHE

50 µG
Catalog # CAYM33300-50
Supplier:   CAYMAN
(±)-7-HDoHE
Catalog # CAYM33300-50
Supplier:  CAYMAN
CAS Number:  90780-55-5
(±)-7-HDoHE

50 µG

CAS Number:  90780-55-5
Price
259,35 €
Log in for availability
1 * 50 µG
Product Details & Documents
  • CAS Number:90780-55-5
  • Formula:C₂₂H₃₂O₃
  • UN:1170
  • ADR:3,II
Specifications

Specifications

  • Pk:
    50 µG

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.