Order Entry
Slovakia
Orders LinkContactUsLinkComponent
(±)-7-HDoHE

(±)-7-HDoHE

25 µG
Catalog # CAYM33300-25
Supplier:   CAYMAN
(±)-7-HDoHE
Catalog # CAYM33300-25
Supplier:  CAYMAN
CAS Number:  90780-55-5
(±)-7-HDoHE

25 µG

CAS Number:  90780-55-5
Price
138,10 €
Log in for availability
1 * 25 µG
Product Details & Documents
  • CAS Number:90780-55-5
  • Formula:C₂₂H₃₂O₃
  • UN:1170
  • ADR:3,II
Specifications

Specifications

  • Pk:
    25 µG

Specifications