Order Entry
Slovakia
ContactUsLinkComponent
Recombinant protein, acads( aah25963.1, 1 a.a. - 412 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000035-P01
Dodávateľ:  ABNOVA
Recombinant protein, acads( aah25963.1, 1 a.a. - 412 a.a.) full-length recombinant protein with gst tag. 1 * 10 µG
Catalog # ABNOH00000035-P01
Dodávateľ:  ABNOVA
Súvisiace informácie
Frequently Bought Together