MICROSART FILTER 100ML 0.45#m RC white (w/o grid) 47mm, sterile, TRAY version 1 * 24 items
Catalog # 516-0397
Dodávateľ: Sartorius
MICROSART FILTER 100ML 0.45#m RC white (w/o grid) 47mm, sterile, TRAY version 1 * 24 items
Catalog # 516-0397
Dodávateľ: Sartorius
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodávateľ: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Dodávateľ: VWR Chemicals
Sealing film for manual application, Parafilm® M
Dodávateľ: HEATHROW
Acetone ≥99%, TECHNICAL
Dodávateľ: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodávateľ: VWR Chemicals
VWR®, Nitrile Gloves
Dodávateľ: VWR Collection
VWR® Facial Tissues
Dodávateľ: VWR Collection
VWR®, Transfer Pipettes, High Performance
Dodávateľ: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Dodávateľ: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Dodávateľ: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Dodávateľ: VWR Collection