D-(-)-Sorbitol ≥98%
Glass bottle for solids, 500 g
Catalog # ABCRAB208062.0500
Supplier: abcr
Jump to:
Product Details & Documents
Purity: 98% CAS No.: 50-70-4 Molecular Formula: C6H14O6 Formula Weight: 182.17 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White Odour: Odourless Melting Point: 98-100° Solubility: Soluble in water, methanol, acetone, DMF Beilstein Reference: 1721899 Merck Reference: 14,8725 MDL No.: MFCD00004708
Synonyms: (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
- CAS Number:50-70-4
- Formula:HOH₂C(CH(OH))₄CH₂OH
- MDL Number:MFCD00004708
- Molecular Weight:182.17 g/mol
- Merck Index:14,8725
- Boiling Pt:295 °C (1013 hPa)
- Melting Pt:98…100 °C
- Density:1.524 g/cm³ (20 °C)
- Flash Pt:>100 °C
- Storage Temperature:Ambient
- EINECS:200-061-5
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



