Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Mouse IL-2 ELISPOT Kit

Mouse IL-2 ELISPOT Kit

Mouse IL-2 ELISPOT kit, 96 tests
Catalog # ABCAAB64031-96
Supplier:   ABCAM
Mouse IL-2 ELISPOT Kit
Catalog # ABCAAB64031-96
Supplier:  ABCAM
CAS Number:  
Mouse IL-2 ELISPOT Kit
Mouse IL-2 ELISPOT Kit

Mouse IL-2 ELISPOT kit, 96 tests

CAS Number:  
Price
550,35 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Mouse IL-2 ELISPOT kit is a ELISPOT kit for the sensitive detection of Mouse IL-2 production in a single cell suspension.

Specifications

Specifications

  • Description:
    Mouse IL-2 ELISPOT kit
  • Tests per kit:
    96 tests
  • Cat. No.:
    ABCAAB64031-96
  • Sample type:
    Suspension cells
  • Form:
    Pre-coated
  • Species reactivity:
    Mouse
  • Storage temperature:
    4 ℃
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO
  • Antigen symbol:
    IL-2

Specifications