Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovakia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HALFMASK X-PLORE 3300 - A1B1E1K1HgP3 1 * 1 items
Catalog # DRAER57794
Supplier:
Dräger
Expand image
HALFMASK X-PLORE 3300 - A1B1E1K1HgP3 1 * 1 items
Catalog # DRAER57794
Supplier:
Dräger
Frequently Bought Together
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
Gloves, nitrile/neoprene, Microflex® 93-260
Supplier: Ansell
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Nitrile Extra Light, Nitrile Gloves
Supplier: VWR Collection
VWR®, Latex Gloves
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Labels, Hazardous Substance, GHS Labels
Supplier: VWR Collection
Beakers, tall form
Supplier: DWK LIFE SCIENCES