Order Entry
Slovakia
Orders LinkContactUsLinkComponent
Bismuth(III) nitrate basic, Sigma-Aldrich®

Bismuth(III) nitrate basic, Sigma-Aldrich®

Glass bottle for solids, 500 g
Catalog # SIALB0426-500G
Oxidizing
Irritant
Supplier:   Merck
Bismuth(III) nitrate basic, Sigma-Aldrich®
Catalog # SIALB0426-500G
Supplier:  Merck
CAS Number:  1304-85-4
Bismuth(III) nitrate basic, Sigma-Aldrich®

Glass bottle for solids, 500 g

CAS Number:  1304-85-4
Price
767,37 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Bismuth(III) nitrate alkaline, Bismuth hydroxide nitrate oxide, Bismuth(III) hydroxide nitrate oxide, Bismuth subnitrate
  • CAS Number:1304-85-4
  • Formula:Bi₅O(OH)₉(NO₃)₄
  • MDL Number:MFCD00064844
  • Molecular Weight:1461.99 g/mol
  • Density:4.928 g/cm³ (20 °C)
  • EINECS:215-136-8
  • UN:1477
  • ADR:5.1,II
  •  :M-Clarity™ quality level = MQ200
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications