Order Entry
Slovakia
Orders LinkContactUsLinkComponent
 

Sample vials, amber, without caps, KIMBLE®

12,00 ml, 19×65 mm, Amber, No, 15-425
Catalog # SCERSP60815-1965
Supplier:   SCHERF PRAZISION
Sample vials, amber, without caps, KIMBLE®
Catalog # SCERSP60815-1965
Supplier:  SCHERF PRAZISION
Sample vials, amber, without caps, KIMBLE®

12,00 ml, 19×65 mm, Amber, No, 15-425

Price
245,79 €
Log in for availability
1 * 200 items
Product Details & Documents

These amber screw thread sample vials without closures are manufactured from 51 expansion borosilicate glass.

 : Meets USP Type I

Specifications
  • Cap included:
    No
  • Cap Size:
    15-425
  • Ø×H:
    19×65 mm
  • Capacity:
    12,00 ml
  • Colour:
    Amber