Adapter piece for chemistry diaphragm pump NT, for a PTFE tubing adapter, In short design 1 * 1 items
Catalog # VACU20636274
Supplier: VACUUBRAND
Adapter piece for chemistry diaphragm pump NT, for a PTFE tubing adapter, In short design 1 * 1 items
Catalog # VACU20636274
Supplier: VACUUBRAND
Frequently Bought Together
Accessories for accu-jet® pro
Supplier: BRAND
Accessories for Research® Plus pipettes
Supplier: EPPENDORF
Accessories for electronic dispensers, Multipette E3/E3x
Supplier: EPPENDORF
Seals for threaded connection caps, BOLA
Supplier: Bohlender
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Mini vortex mixer
Supplier: VWR Collection
Lithium Polymer Rechargable Battery
Supplier: EPPENDORF
Acetonitrile, LiChrosolv®, hypergrade for LC-MS, Supelco®
Supplier: Merck
VWR®, Accessories for Mechanical Pipettes
Supplier: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Cover gasket 1 * 1 items
Supplier: VWR Collection
Accessories for HandyStep® electronic
Supplier: BRAND
Accessories for pipetus® / pipetus®-standard / pipetus®-junior
Supplier: HIRSCHMANN
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals