Order Entry
Slovakia
ContactUsLinkComponent
Filter, disc diameter 240mm, material glass microfiber, grade MGF, binder-free, particle retention is 0.7 µm, suitable for TCLP anal ysis and the retention of very fine particles. 1 * 25 items
  SARTFT-3-1105-240
 :  Sartorius
Filter, disc diameter 240mm, material glass microfiber, grade MGF, binder-free, particle retention is 0.7 µm, suitable for TCLP anal ysis and the retention of very fine particles. 1 * 25 items
  SARTFT-3-1105-240
 :  Sartorius
Frequently Bought Together